Learn More
Waters Corp ACQUITY UPLC CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 1 mm X 50 mm, 3/pk
Supplier:  Waters Corp 176002150
Encompass_Preferred
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-117