Learn More
Waters Corp ACQUITY UPLC CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 1 mm X 150 mm, 1/pk
Catalog No. 50791372 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-372 Supplier Waters Corp Supplier No. 186005348
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.