Learn More
Waters Corp Waters ACQUITY UPLC CSH Fluoro-Phenyl Column, 1.7 μm, 3 mm × 150 mm, 130 Å
Catalog No. 50791376 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY CSH Fluoro-Phenyl column is designed to maximize selectivity differences, offering method development freedom, especially with low pH mobile phases. The non-encapped sorbent enhances the retention of acidic compounds.
  • Maximizes selectivity differences in separations.
  • Offers method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • Particle size of 1.7 μm.
  • Inner diameter of 3 mm and length of 150 mm.
  • Pore size of 130 Å.
  • Operates effectively within a pH range of 1 to 8.
  • Withstands a maximum pressure of 18000 psi (1240 Bar).
  • Utilizes PFP/Fluoro-Phenyl chemistry.
  • Based on CSH particle technology.
  • Compatible with UPLC and UHPLC systems.
Catalog No. 50-791-376 Supplier Waters Corp Supplier No. 186005357
The ACQUITY CSH Fluoro-Phenyl column is designed to maximize selectivity differences, offering method development freedom, especially with low pH mobile phases. The non-encapped sorbent enhances the retention of acidic compounds.
  • Maximizes selectivity differences in separations.
  • Offers method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • Particle size of 1.7 μm.
  • Inner diameter of 3 mm and length of 150 mm.
  • Pore size of 130 Å.
  • Operates effectively within a pH range of 1 to 8.
  • Withstands a maximum pressure of 18000 psi (1240 Bar).
  • Utilizes PFP/Fluoro-Phenyl chemistry.
  • Based on CSH particle technology.
  • Compatible with UPLC and UHPLC systems.