Learn More
Waters Corp Waters UPLC CSH Fluoro-Phenyl Column, 1.7 μm, 130Å, 1 mm x 100 mm
Supplier:  Waters Corp 186005347
Encompass_Preferred
This Fluoro-Phenyl column is designed to maximize selectivity differences, providing maximum method development freedom, especially when using low pH mobile phases. Its non-encapped sorbent offers enhanced retention of acidic compounds.
  • Designed to maximize selectivity differences
  • Provides maximum method development freedom
  • Especially useful with low pH mobile phases
  • Non-encapped sorbent provides enhanced retention of acidic compounds
Catalog No. 50-791-371