Learn More
Waters Corp Waters ACQUITY UPLC CSH Fluoro-Phenyl Column, 1.7 μm, 2.1 x 30 mm
Supplier:  Waters Corp 176002151
Encompass_Preferred
ACQUITY UPLC CSH Fluoro-Phenyl columns address the challenge of maintaining acceptable batch-to-batch reproducibility in stationary phases with large selectivity differences. These columns utilize CSH Technology bonding process to achieve exceptional reproducibility and sustained selectivity across particle sizes.
  • Offers exceptional batch-to-batch reproducibility due to the CSH Technology bonding process.
  • Provides sustained selectivity across particle sizes.
  • Pentafluorophenyl (PFP) ligands provide multiple retention mechanisms including hydrogen bonding, dipole-dipole interaction, aromatic (π–π), and hydrophobic (reversed-phase) retention, useful for separating isomeric, aromatic, or halogenated compounds.
Catalog No. 50-791-118