Learn More
Waters Corp ACQUITY UPLC CSH Fluoro-Phenyl Column, 130Å, 1.7 µm, 3 mm X 50 mm, 3/pk
Catalog No. 50791122 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.
Catalog No. 50-791-122 Supplier Waters Corp Supplier No. 176002156
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds.