Learn More
Waters Corp Waters ACQUITY UPLC CSH Fluoro-Phenyl Column, 130Å, 1.7 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 176002154
Encompass_Preferred
The ACQUITY UPLC CSH Fluoro-Phenyl Column offers enhanced selectivity, especially with low pH mobile phases, utilizing Charged Surface Hybrid (CSH) Technology and multiple retention mechanisms.
  • Maximizes selectivity differences, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
  • Offers outstanding selectivity for positional isomers, halogenated, and polar compounds.
  • Combines multiple retention mechanisms: dipole-dipole, hydrogen-bonding, aromatic (pi-pi), and hydrophobic interactions.
  • Developed using Charged Surface Hybrid (CSH) Technology.
  • Produced in a cGMP, ISO 9001-certified facility.
  • Each batch tested chromatographically for repeatable performance.
Catalog No. 50-791-120