Learn More
Waters Corp Waters Corporation Column, Phenyl-Hexyl, 1.7 μm, 3 mm × 50 mm
Catalog No. 50791424 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH Phenyl-Hexyl columns are designed to offer alternative analyte selectivity and serve as a valuable tool for method development. The trifunctionally bonded C6 phenyl ligand provides robust, low-bleed sorbents that selectively retain polyaromatic compounds through pi-pi interactions.
  • Trifunctionally bonded C6 phenyl ligand
  • Robust, low bleed sorbents
  • Selectively retains polyaromatic compounds through pi-pi interactions
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range from 1 to 11
  • Maximum pressure of 18000 psi (1240 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 130 Å pore size
  • Compatible with UPLC and UHPLC systems
Catalog No. 50-791-424 Supplier Waters Corp Supplier No. 186005410
ACQUITY CSH Phenyl-Hexyl columns are designed to offer alternative analyte selectivity and serve as a valuable tool for method development. The trifunctionally bonded C6 phenyl ligand provides robust, low-bleed sorbents that selectively retain polyaromatic compounds through pi-pi interactions.
  • Trifunctionally bonded C6 phenyl ligand
  • Robust, low bleed sorbents
  • Selectively retains polyaromatic compounds through pi-pi interactions
  • Reversed phase separation mode
  • Hybrid particle substrate
  • pH range from 1 to 11
  • Maximum pressure of 18000 psi (1240 Bar)
  • Endcapped
  • Low silanol activity
  • Spherical particle shape
  • 130 Å pore size
  • Compatible with UPLC and UHPLC systems