Learn More
Waters Corp ACQUITY CSH Phenyl-Hexyl Column, 1.7 μm, 130 Å, 3.0 x 30 mm
Catalog No. 50791423 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC CSH Phenyl-Hexyl Column offers complementary selectivity for polyaromatic compounds. Built on the Charged Surface Hybrid (CSH) particle platform, it delivers exceptional peak shape under low and high pH conditions.
  • Based on the Charged Surface Hybrid (CSH) particle platform.
  • Offers complementary selectivity, especially for polyaromatic compounds.
  • Provides exceptional peak shape for basic compounds under low and high pH conditions.
  • Features a trifunctional C6 Phenyl ligand, with a 14% carbon load.
  • Operates within a pH range of 1 to 11.
  • Temperature limit of 80°C at low pH and 45°C at high pH.
  • Pore diameter of 130 Å and surface area of 185 m2/g.
  • Effective for analyzing basic drugs and characterizing compounds like clozapine.
Catalog No. 50-791-423 Supplier Waters Corp Supplier No. 186005409
The ACQUITY UPLC CSH Phenyl-Hexyl Column offers complementary selectivity for polyaromatic compounds. Built on the Charged Surface Hybrid (CSH) particle platform, it delivers exceptional peak shape under low and high pH conditions.
  • Based on the Charged Surface Hybrid (CSH) particle platform.
  • Offers complementary selectivity, especially for polyaromatic compounds.
  • Provides exceptional peak shape for basic compounds under low and high pH conditions.
  • Features a trifunctional C6 Phenyl ligand, with a 14% carbon load.
  • Operates within a pH range of 1 to 11.
  • Temperature limit of 80°C at low pH and 45°C at high pH.
  • Pore diameter of 130 Å and surface area of 185 m2/g.
  • Effective for analyzing basic drugs and characterizing compounds like clozapine.