Learn More
Waters Corp Waters ACQUITY UPLC CSH Phenyl-Hexyl Column, 1.7 μm Particle, 130 Å Pore, 3.0 × 30 mm
Catalog No. 50791131 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC CSH Phenyl-Hexyl Columns are designed to provide superior peak shape for basic analytes when used with formic acid or other low-ionic-strength, acidic mobile phases.
  • Provides superior peak shape for basic analytes.
  • Achieved through a controlled, low-level, positive surface charge.
  • Bonded to ethylene-bridged hybrid (BEH) particles.
  • Eliminates the need for ion-pairing reagents.
  • Rugged design.
Catalog No. 50-791-131 Supplier Waters Corp Supplier No. 176002166
ACQUITY UPLC CSH Phenyl-Hexyl Columns are designed to provide superior peak shape for basic analytes when used with formic acid or other low-ionic-strength, acidic mobile phases.
  • Provides superior peak shape for basic analytes.
  • Achieved through a controlled, low-level, positive surface charge.
  • Bonded to ethylene-bridged hybrid (BEH) particles.
  • Eliminates the need for ion-pairing reagents.
  • Rugged design.