Learn More
Waters Corp ACQUITY UPLC CSH Phenyl-Hexyl Column, 130Å, 1.7 µm, 3 mm X 50 mm, 3/pk
Supplier:  Waters Corp 176002167
Encompass_Preferred
ACQUITY CSH Phenyl-Hexyl columns are used to provide alternative analyte selectivity and are a valuable tool for method development. The trifunctionally bonded C6 phenyl ligand is a robust, low bleed sorbents that selectively retains polyaromotic compounds through pi-pi interactions.
Catalog No. 50-791-132