Learn More
Waters Corp WATERS Column, Phenyl-Hexyl, 1.7 μm, 1x50 mm
Catalog No. 50791420 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY CSH Phenyl-Hexyl columns are a valuable tool for method development, providing alternative analyte selectivity. These columns feature a robust, low bleed C6 phenyl ligand that selectively retains polyaromatic compounds through pi-pi interactions, ensuring diverse selectivities, excellent peak shape, and reduced column bleed.
  • Provides alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl ligand selectively retains polyaromatic compounds.
  • Offers diverse selectivities, excellent peak shape, and reduced column bleed.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Chromatographically tested for repeatable performance.
  • Wide pH range of 1–11 pH.
Catalog No. 50-791-420 Supplier Waters Corp Supplier No. 186005404
ACQUITY CSH Phenyl-Hexyl columns are a valuable tool for method development, providing alternative analyte selectivity. These columns feature a robust, low bleed C6 phenyl ligand that selectively retains polyaromatic compounds through pi-pi interactions, ensuring diverse selectivities, excellent peak shape, and reduced column bleed.
  • Provides alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl ligand selectively retains polyaromatic compounds.
  • Offers diverse selectivities, excellent peak shape, and reduced column bleed.
  • Manufactured in a cGMP, ISO 9001-certified facility.
  • Chromatographically tested for repeatable performance.
  • Wide pH range of 1–11 pH.