Learn More
Waters Corp ACQUITY UPLC CSH Phenyl-Hexyl Method Validation Kit, 130Å, 1.7 µm, 2.1 mm X 50 mm, 3/pk
Catalog No. 50791536 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit consists of three ACQUITY CSH Phenyl-Hexyl columns packed with three different batches of sorbent.
Catalog No. 50-791-536 Supplier Waters Corp Supplier No. 186005579
This kit consists of three ACQUITY CSH Phenyl-Hexyl columns packed with three different batches of sorbent.