Learn More
Waters Corp Waters Corporation Assembly, Cuvette Holder, 2 Cuvettes, ACQUITY FLR
Catalog No. 50791686 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This assembly includes a cuvette holder and two cuvettes, specifically designed for use with ACQUITY FLR systems. It functions as a cuvette flow cell, enabling the scanning and plotting of excitation and emission wavelength ranges, crucial for maintaining and enhancing system performance.
  • Includes a cuvette holder and two cuvettes.
  • Designed for ACQUITY FLR systems.
  • Scans and plots excitation and emission wavelength ranges.
  • Enhances performance of ACQUITY FLR systems.
Catalog No. 50-791-686 Supplier Waters Corp Supplier No. 205000500
Add to Cart
Add to Cart
Request a Quote
This assembly includes a cuvette holder and two cuvettes, specifically designed for use with ACQUITY FLR systems. It functions as a cuvette flow cell, enabling the scanning and plotting of excitation and emission wavelength ranges, crucial for maintaining and enhancing system performance.
  • Includes a cuvette holder and two cuvettes.
  • Designed for ACQUITY FLR systems.
  • Scans and plots excitation and emission wavelength ranges.
  • Enhances performance of ACQUITY FLR systems.