Learn More
Waters Corp Waters Upgrade Kit, ACQUITY UPLC H-Class Bio, Hexane/THF Mobile Phase
Catalog No. 50466541 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This upgrade kit is designed for ACQUITY UPLC H-Class Series and H-Class Bio Series Systems. It enables these systems to safely and effectively utilize hexane and tetrahydrofuran (THF) in their solvent systems, expanding the range of applications and analyses that can be performed with these systems.
  • Upgrades systems to safely use hexane and THF in solvent systems.
  • Expands the range of applications and analyses for UPLC systems.
  • Includes replacement components for quaternary solvent manager modifications.
  • Provides O-ring for drain cup replacement.
  • Offers hexane/THF compatible i2Valve cartridge for primary check valve.
  • Supplies convoluted tubing for waste lines.
  • Includes adapter fitting for seal-wash pump outlet.
  • Contains back-pressure restrictor for UV detectors.
  • Facilitates extension of degasser vent line for vapor routing.
Catalog No. 50-466-541 Supplier Waters Corp Supplier No. 205000800
This upgrade kit is designed for ACQUITY UPLC H-Class Series and H-Class Bio Series Systems. It enables these systems to safely and effectively utilize hexane and tetrahydrofuran (THF) in their solvent systems, expanding the range of applications and analyses that can be performed with these systems.
  • Upgrades systems to safely use hexane and THF in solvent systems.
  • Expands the range of applications and analyses for UPLC systems.
  • Includes replacement components for quaternary solvent manager modifications.
  • Provides O-ring for drain cup replacement.
  • Offers hexane/THF compatible i2Valve cartridge for primary check valve.
  • Supplies convoluted tubing for waste lines.
  • Includes adapter fitting for seal-wash pump outlet.
  • Contains back-pressure restrictor for UV detectors.
  • Facilitates extension of degasser vent line for vapor routing.