Learn More
Waters Corp Performance Maintenance Kit, ACQUITY H-Class Bio-QSM, UPLC, Annual
Catalog No. 502391217 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit includes all the quality parts needed to maintain the instrument's peak performance annually. The kit contains plungers, check valves, seals, and filters.
  • Performance maintenance type
  • System type: UPLC
  • Instrument: ACQUITY Arc Premier System
  • Replacement schedule: Annual
  • Component: ACQUITY UPLC QSM
  • Units per package: 1 pk
Catalog No. 50-239-1217 Supplier Waters Corp Supplier No. 201000244
The Performance Maintenance Kit includes all the quality parts needed to maintain the instrument's peak performance annually. The kit contains plungers, check valves, seals, and filters.
  • Performance maintenance type
  • System type: UPLC
  • Instrument: ACQUITY Arc Premier System
  • Replacement schedule: Annual
  • Component: ACQUITY UPLC QSM
  • Units per package: 1 pk