Learn More
Waters Corp Performance Maintenance Kit, ACQUITY H-Class Bio QSM, UPLC, Annual, w/PCV
Catalog No. 502390707 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit for the H-Class Bio Quaternary Solvent Manager with Passive Check Valves includes all necessary Waters Quality Parts to ensure the instrument operates at peak performance annually. It contains essential components such as the mixer, plungers, check valves, seals, and filters.
  • For UPLC system type.
  • Compatible with ACQUITY UPLC H-Class System and ACQUITY Premier System.
  • Designed for annual replacement.
  • Kit includes mixer, plungers, check valves, seals, and filters.
  • Contains one package.
Catalog No. 50-239-0707 Supplier Waters Corp Supplier No. 201000305
This Performance Maintenance Kit for the H-Class Bio Quaternary Solvent Manager with Passive Check Valves includes all necessary Waters Quality Parts to ensure the instrument operates at peak performance annually. It contains essential components such as the mixer, plungers, check valves, seals, and filters.
  • For UPLC system type.
  • Compatible with ACQUITY UPLC H-Class System and ACQUITY Premier System.
  • Designed for annual replacement.
  • Kit includes mixer, plungers, check valves, seals, and filters.
  • Contains one package.