Learn More
Waters Corp Performance Maintenance Kit, ACQUITY H-Class CM-A / CM-Aux & Bio, Annual
Catalog No. 502390602 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for H-Class CM-A / CM-Aux and Bio contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Designed for UPLC system types.
  • Compatible with ACQUITY Arc Premier System, ACQUITY UPLC H-Class System, ACQUITY Premier, and UPC2 instruments.
  • Includes components for ACQUITY UPLC Column Manager.
  • Contains air filters.
  • Comes with 1 package.
Catalog No. 50-239-0602 Supplier Waters Corp Supplier No. 201000207
The Waters™ Performance Maintenance Kit for H-Class CM-A / CM-Aux and Bio contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Designed for UPLC system types.
  • Compatible with ACQUITY Arc Premier System, ACQUITY UPLC H-Class System, ACQUITY Premier, and UPC2 instruments.
  • Includes components for ACQUITY UPLC Column Manager.
  • Contains air filters.
  • Comes with 1 package.