Learn More
Waters Corp ACQUITY CM-A Tubing Kit, 6 Column, MP35N
Catalog No. 502391957 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit provides MP35N tubing assemblies for implementing CM-A active column managers onto a Bio H-Class PLUS system, supporting a six-column configuration. It also serves as a replacement for all MP35N tubing in this setup and includes the necessary Active Preheater for PLUS systems.
  • Designed for CM-A active column managers
  • Supports a six-column configuration
  • Includes MP35N tubing assemblies
  • Can replace MP35N tubing in compatible configurations
  • Includes an Active Preheater for PLUS systems
  • Compatible with UPLC H-Class, I-Class, and M-Class systems
  • Replacement as required
  • UPLC ASM component included
Catalog No. 50-239-1957 Supplier Waters Corp Supplier No. 205001825
Add to Cart
Add to Cart
Request a Quote
This tubing kit provides MP35N tubing assemblies for implementing CM-A active column managers onto a Bio H-Class PLUS system, supporting a six-column configuration. It also serves as a replacement for all MP35N tubing in this setup and includes the necessary Active Preheater for PLUS systems.
  • Designed for CM-A active column managers
  • Supports a six-column configuration
  • Includes MP35N tubing assemblies
  • Can replace MP35N tubing in compatible configurations
  • Includes an Active Preheater for PLUS systems
  • Compatible with UPLC H-Class, I-Class, and M-Class systems
  • Replacement as required
  • UPLC ASM component included