Learn More
Waters Corp Tubing Kit, ACQUITY H-Class PLUS Bio CM-A, 2 Column, MP35N
Catalog No. 502392941 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit is designed for use with the ACQUITY H-Class PLUS Bio CM-A system. It contains MP35N tubing assemblies to implement a CM-A active column manager onto a Bio ACQUITY H-Class PLUS, supporting a two-column configuration. This kit can also be used to replace existing MP35N tubing in this configuration and includes the relevant Active Preheater for a PLUS system.
  • Tubing, performance maintenance, and upgrade kit
  • UPLC system compatibility
  • Compatible with ACQUITY UPLC H-Class, I-Class, and M-Class systems
  • Replacement as required
  • ACQUITY UPLC ASM component
  • Constructed with MP35N tubing
  • Implements a CM-A active column manager, supports a two-column configuration, and replaces MP35N tubing
  • Includes relevant active preheater for a PLUS system
Catalog No. 50-239-2941 Supplier Waters Corp Supplier No. 205001823
This tubing kit is designed for use with the ACQUITY H-Class PLUS Bio CM-A system. It contains MP35N tubing assemblies to implement a CM-A active column manager onto a Bio ACQUITY H-Class PLUS, supporting a two-column configuration. This kit can also be used to replace existing MP35N tubing in this configuration and includes the relevant Active Preheater for a PLUS system.
  • Tubing, performance maintenance, and upgrade kit
  • UPLC system compatibility
  • Compatible with ACQUITY UPLC H-Class, I-Class, and M-Class systems
  • Replacement as required
  • ACQUITY UPLC ASM component
  • Constructed with MP35N tubing
  • Implements a CM-A active column manager, supports a two-column configuration, and replaces MP35N tubing
  • Includes relevant active preheater for a PLUS system