Learn More
Waters Corp Performance Maintenance Kit, for ACQUITY H-Class PLUS Bio Sample Manager FTN, Annual
Catalog No. 502392754 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit is designed for the ACQUITY H-Class PLUS Bio Sample Manager – Flow Through Needle. This kit contains all the necessary Waters Quality Parts™ to keep the instrument operating at peak performance on an annual basis.
  • Includes essential quality parts for instrument operation.
  • Designed for annual maintenance.
  • Kit includes syringe, standard needle, needle seat, injection cartridge, and filters.
  • Compatible with UPLC system type.
  • For ACQUITY instruments.
Catalog No. 50-239-2754 Supplier Waters Corp Supplier No. 201000315
The Waters Performance Maintenance Kit is designed for the ACQUITY H-Class PLUS Bio Sample Manager – Flow Through Needle. This kit contains all the necessary Waters Quality Parts™ to keep the instrument operating at peak performance on an annual basis.
  • Includes essential quality parts for instrument operation.
  • Designed for annual maintenance.
  • Kit includes syringe, standard needle, needle seat, injection cartridge, and filters.
  • Compatible with UPLC system type.
  • For ACQUITY instruments.