Learn More
Waters Corp Tubing Kit, Stainless Steel, UPLC, 6 Column
Catalog No. 502391799 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit contains stainless steel tubing assemblies for implementing CM-A active column managers onto an ACQUITY H-Class PLUS system. It supports a six-column configuration and can also be used to replace existing stainless steel tubing in this setup. The kit includes the relevant Active Preheater for a PLUS system.
  • Designed for UPLC system configurations
  • Suitable for Arc Premier, UPLC H-Class, and Patrol instruments
  • Replaces stainless steel tubing in six-column setups
  • Includes an active preheater for PLUS systems
Catalog No. 50-239-1799 Supplier Waters Corp Supplier No. 205001822
Add to Cart
Add to Cart
This tubing kit contains stainless steel tubing assemblies for implementing CM-A active column managers onto an ACQUITY H-Class PLUS system. It supports a six-column configuration and can also be used to replace existing stainless steel tubing in this setup. The kit includes the relevant Active Preheater for a PLUS system.
  • Designed for UPLC system configurations
  • Suitable for Arc Premier, UPLC H-Class, and Patrol instruments
  • Replaces stainless steel tubing in six-column setups
  • Includes an active preheater for PLUS systems