Learn More
Waters Corp Performance Maintenance Kit, ACQUITY H-Class QSM, Passive Check Valves, Annual
Catalog No. 502391185 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Performance Maintenance Kit for the H-Class Quaternary Solvent Manager with Passive Check Valves is designed to ensure your instrument operates at peak performance on an annual basis. This kit includes all necessary Waters Quality Parts.
  • Performance maintenance kits
  • UPLC system type
  • For ACQUITY UPLC H-Class system
  • Annual replacement schedule
  • ACQUITY UPLC QSM component
  • One unit per package
  • Kit includes mixer, plungers, check valves, seals, and filters
Catalog No. 50-239-1185 Supplier Waters Corp Supplier No. 201000304
The Waters Performance Maintenance Kit for the H-Class Quaternary Solvent Manager with Passive Check Valves is designed to ensure your instrument operates at peak performance on an annual basis. This kit includes all necessary Waters Quality Parts.
  • Performance maintenance kits
  • UPLC system type
  • For ACQUITY UPLC H-Class system
  • Annual replacement schedule
  • ACQUITY UPLC QSM component
  • One unit per package
  • Kit includes mixer, plungers, check valves, seals, and filters