Learn More
Waters Corp Waters ACQUITY UPLC HSS Cyano (CN) Column, 1.8 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186005988
Encompass_Preferred
The ACQUITY UPLC HSS Cyano (CN) Column offers ultra-performance general purpose propyl cyano bonded phase chemistry. It delivers superior peak shape for bases, increased retention, and long column lifetimes at low pH. This column is scalable to HPLC and compatible with both reversed-phase and normal-phase techniques.
  • Superior peak shape for bases and increased retention.
  • Extremely long column lifetimes at low pH.
  • Low hydrophobicity and unique selectivity compared to straight-chain alkyl columns.
  • Ultra-stable retention and exceptional peak shape.
  • Reproducibility under low-to mid-pH conditions.
  • Compatible with both reversed-phase and normal-phase techniques.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant.
  • Every column individually tested; performance chromatogram and certificate of batch analysis available on intelligent chip.
Catalog No. 50-179-452