Learn More
Waters Corp Waters UPLC HSS Cyano Column, 100Å, 1.8 μm, 3 mm × 100 mm
Supplier:  Waters Corp 186005993
Encompass_Preferred
The ACQUITY UPLC HSS CN chemistry offers an ultra-performance general purpose propyl cyano bonded phase. It provides superior peak shape for bases, increased retention, and exceptionally long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations, with packing materials specifically designed for UPLC systems.
  • Superior peak shape for bases.
  • Increased retention and extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
  • Packing materials specifically created for UPLC systems.
  • Manufactured in a cGMP, ISO 9001:2000 certified facility using ultra-pure reagents.
  • Offers minimal hydrophobicity and exceptional selectivity compared to straight-chain alkyl columns.
  • Compatible with both normal-phase and reversed-phase methods.
Catalog No. 50-179-457