Learn More
Waters Corp Waters ACQUITY UPLC HSS Cyano Column, 1.8 μm, 2.1 x 50 mm
Catalog No. 50179460 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These ACQUITY UPLC HSS Cyano columns offer increased retention for both polar and non-polar analytes. They feature a thermally treated silica particle for mechanical strength and stability under UPLC system pressures. This Method Validation Kit contains three columns from three different material batches, with a 1.8 μm particle size and 2.1 × 50 mm dimensions.
  • Increased retention for polar and non-polar analytes.
  • Thermally treated silica particle provides mechanical strength and stability under UPLC system pressures.
  • HSS particle technology maximizes selectivity space.
  • Bonded phases support hydrophobic, reversed-phase, dipole-dipole, aromatic, and hydrogen-bonding interactions.
  • Method Validation Kit includes three columns from three batches of material.
  • Particle size: 1.8 μm.
  • Column dimensions: 2.1 × 50 mm.
Catalog No. 50-179-460 Supplier Waters Corp Supplier No. 186005996
These ACQUITY UPLC HSS Cyano columns offer increased retention for both polar and non-polar analytes. They feature a thermally treated silica particle for mechanical strength and stability under UPLC system pressures. This Method Validation Kit contains three columns from three different material batches, with a 1.8 μm particle size and 2.1 × 50 mm dimensions.
  • Increased retention for polar and non-polar analytes.
  • Thermally treated silica particle provides mechanical strength and stability under UPLC system pressures.
  • HSS particle technology maximizes selectivity space.
  • Bonded phases support hydrophobic, reversed-phase, dipole-dipole, aromatic, and hydrogen-bonding interactions.
  • Method Validation Kit includes three columns from three batches of material.
  • Particle size: 1.8 μm.
  • Column dimensions: 2.1 × 50 mm.