Learn More
Waters Corp Waters ACQUITY UPLC HSS PFP Column, 1.8 μm, 100Å, 1 x 100 mm
Catalog No. 50179433 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This non-endcapped silica-based column is designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. These columns are fully scalable and transferable to HPLC separations.
  • Designed for low pH separations
  • Offers alternative selectivity to fully endcapped C18 phases
  • Fully scalable and transferable to HPLC separations
  • Non-endcapped silica-based column
Catalog No. 50-179-433 Supplier Waters Corp Supplier No. 186005962
Add to Cart
Add to Cart
This non-endcapped silica-based column is designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. These columns are fully scalable and transferable to HPLC separations.
  • Designed for low pH separations
  • Offers alternative selectivity to fully endcapped C18 phases
  • Fully scalable and transferable to HPLC separations
  • Non-endcapped silica-based column