Learn More
Waters Corp Waters Column, PFP, 1.8 μm, 2.1 mm X 150 mm, 100 Å
Supplier:  Waters Corp 186005968
Encompass_Preferred
The ACQUITY UPLC HSS PFP column is a non-endcapped, silica-based column designed for low pH separations, offering alternative selectivity to fully endcapped C18 phases. It is fully scalable and transferable to HPLC separations.
  • Features trifunctionally-bonded, non-end-capped pentafluorophenyl (PFP) chemistry for diverse interactions.
  • Optimized for analyzing planar aromatic compounds, positional isomers, halogenated chemicals, and polar analytes.
  • Provides industry-leading repeatability, peak form, and scalability for PFP stationary phases.
  • Offers remarkable batch-to-batch repeatability and sustained selectivity across UPLC and HPLC particle sizes.
  • Chromatographically tested to ensure exceptional, repeatable performance with acidic, basic, and neutral analytes.
  • Delivers outstanding peak form and retention for beta-blocker separation.
Catalog No. 50-179-437