Learn More
Waters Corp Waters Corporation Column, PFP/Fluoro-Phenyl, 1.8 μm, 2.1 mm × 50 mm, 100Å
Catalog No. NC0257439 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This non-endcapped silica-based column is designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. It is fully scalable and transferable to HPLC separations.
  • Non-endcapped, silica-based column for low pH separations.
  • Fully scalable and transferable to HPLC separations.
  • Trifunctionally bonded for various aromatic and polar analytes.
  • Utilizes multiple interaction types for molecule retention.
  • Offers industry-leading reproducibility, peak shape, and scalability for PFP stationary phases.
  • Remarkable batch-to-batch reproducibility and sustained selectivity.
  • Chromatographically tested with acidic, basic, and neutral analytes for consistent performance.
  • Manufactured in a cGMP, ISO 9001 certified facility with ultra-pure reagents.
Catalog No. NC0257439 Supplier Waters Corp Supplier No. 186005965
This non-endcapped silica-based column is designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. It is fully scalable and transferable to HPLC separations.
  • Non-endcapped, silica-based column for low pH separations.
  • Fully scalable and transferable to HPLC separations.
  • Trifunctionally bonded for various aromatic and polar analytes.
  • Utilizes multiple interaction types for molecule retention.
  • Offers industry-leading reproducibility, peak shape, and scalability for PFP stationary phases.
  • Remarkable batch-to-batch reproducibility and sustained selectivity.
  • Chromatographically tested with acidic, basic, and neutral analytes for consistent performance.
  • Manufactured in a cGMP, ISO 9001 certified facility with ultra-pure reagents.