Learn More
Waters Corp Waters ACQUITY UPLC HSS PFP Column, 1.8 μm, 3 mm, 100 mm, 100 Å
Catalog No. 50179441 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC HSS PFP column is a non-endcapped, silica-based column for low pH separations, offering alternative selectivity to fully endcapped C18 phases. It is scalable and transferable to HPLC separations, separating polar analytes, halogenated chemicals, isomers, and planar aromatic compounds through hydrophobic, dipole-dipole, hydrogen-bonding, and aromatic (pi-pi) interactions.
  • Non-endcapped silica-based column for low pH separations.
  • Alternative selectivity to fully endcapped C18 phases.
  • Scalable and transferable to HPLC separations.
  • Combines multiple retention mechanisms (hydrophobic, dipole-dipole, hydrogen-bonding, pi-pi interactions).
  • Superior peak form and retention for beta-blocker separation.
  • Excellent batch-to-batch repeatability and consistent selectivity.
  • eCord enabled.
Catalog No. 50-179-441 Supplier Waters Corp Supplier No. 186005972
The ACQUITY UPLC HSS PFP column is a non-endcapped, silica-based column for low pH separations, offering alternative selectivity to fully endcapped C18 phases. It is scalable and transferable to HPLC separations, separating polar analytes, halogenated chemicals, isomers, and planar aromatic compounds through hydrophobic, dipole-dipole, hydrogen-bonding, and aromatic (pi-pi) interactions.
  • Non-endcapped silica-based column for low pH separations.
  • Alternative selectivity to fully endcapped C18 phases.
  • Scalable and transferable to HPLC separations.
  • Combines multiple retention mechanisms (hydrophobic, dipole-dipole, hydrogen-bonding, pi-pi interactions).
  • Superior peak form and retention for beta-blocker separation.
  • Excellent batch-to-batch repeatability and consistent selectivity.
  • eCord enabled.