Learn More
Waters Corp ACQUITY UPLC HSS PFP Column, 100Å, 1.8 µm, 3 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186005970
Encompass_Preferred
ACQUITY UPLC HSS PFP is a non-endcapped silica-based column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. ACQUITY HSS PFP columns are fully scalable and transferable to HPLC separations.
Catalog No. 50-179-439