Learn More
Waters Corp Waters XSelect HSS PFP Method Transfer Kit, 100Å, 1.8 to 5 μm
Supplier:  Waters Corp 186005979
Encompass_Preferred
This method transfer kit is designed for low pH separations, offering alternative selectivity compared to a fully endcapped, high coverage C18 phase. The columns facilitated by this kit are fully scalable and transferable to HPLC separations.
  • PFP/Fluoro-Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range: 2 to 8 pH
  • Non-endcapped
  • PFP bonding technology
  • High silanol activity
  • Spherical particle shape
  • 1.8 to 5 μm particle size
  • 100 Å pore size
  • 230 surface area
  • HSS particle technology
  • L43 USP classification
  • 7 % carbon load
Catalog No. 50-179-444