Learn More
Waters Corp Rebuild Kit, ACQUITY I-Class Accumulator Pumphead
Catalog No. 502391179 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit provides the essential components required to maintain the performance of an ACQUITY I-Class Accumulator Pumphead. It ensures proper functioning and extends the lifespan of the system.
  • Contains the plunger, seals, and check valve.
  • Designed for UPLC systems.
  • Provides components for as-required replacement.
  • Includes one package of components.
Catalog No. 50-239-1179 Supplier Waters Corp Supplier No. 700010584
This rebuild kit provides the essential components required to maintain the performance of an ACQUITY I-Class Accumulator Pumphead. It ensures proper functioning and extends the lifespan of the system.
  • Contains the plunger, seals, and check valve.
  • Designed for UPLC systems.
  • Provides components for as-required replacement.
  • Includes one package of components.