Learn More
Waters Corp Performance Maintenance Kit, ACQUITY I-Class PLUS Sample Manager, Flow Through Needle
Catalog No. 502392766 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the ACQUITY I-Class PLUS Sample Manager – Flow Through Needle contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. The kit includes a syringe, standard needle, needle seat, injection cartridge, and filters.
  • Maintains ACQUITY I-Class PLUS Sample Manager – Flow Through Needle
  • Ensures peak instrument performance
  • Includes syringe, standard needle, needle seat, injection cartridge, and filters
  • Recommended for annual replacement
Catalog No. 50-239-2766 Supplier Waters Corp Supplier No. 201000316
The Waters™ Performance Maintenance Kit for the ACQUITY I-Class PLUS Sample Manager – Flow Through Needle contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. The kit includes a syringe, standard needle, needle seat, injection cartridge, and filters.
  • Maintains ACQUITY I-Class PLUS Sample Manager – Flow Through Needle
  • Ensures peak instrument performance
  • Includes syringe, standard needle, needle seat, injection cartridge, and filters
  • Recommended for annual replacement