Learn More
Waters Corp Waters Rebuild Kit, ACQUITY I-Class, Primary Pumphead, UPLC System
Catalog No. 502039804 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild an ACQUITY I-Class Primary Pumphead.
  • Contains plunger, seals, and check valve.
  • Designed for ACQUITY I-Class Primary Pumphead.
  • Compatible with ACQUITY UPLC I-Class System and ACQUITY instruments.
  • Used with ACQUITY UPLC BSM component.
  • Replacement schedule is as required.
  • One unit per package.
Catalog No. 50-203-9804 Supplier Waters Corp Supplier No. 700010583
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild an ACQUITY I-Class Primary Pumphead.
  • Contains plunger, seals, and check valve.
  • Designed for ACQUITY I-Class Primary Pumphead.
  • Compatible with ACQUITY UPLC I-Class System and ACQUITY instruments.
  • Used with ACQUITY UPLC BSM component.
  • Replacement schedule is as required.
  • One unit per package.