Learn More
Waters Corp Performance Maintenance Kit, ACQUITY I-Class Sample Manager -FL, UPLC System, Annual
Catalog No. 50466534 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed to ensure the optimal operation of the ACQUITY I-Class Sample Manager –Fixed Loop. It provides all the necessary quality parts for annual preventative maintenance, helping to keep your instrument operating at peak performance.
  • Includes syringe, standard needle, and injection cartridge.
  • Contains a 10μL loop and filters.
  • Designed for UPLC system types.
  • Recommended for annual replacement.
Catalog No. 50-466-534 Supplier Waters Corp Supplier No. 201000258
This Performance Maintenance Kit is designed to ensure the optimal operation of the ACQUITY I-Class Sample Manager –Fixed Loop. It provides all the necessary quality parts for annual preventative maintenance, helping to keep your instrument operating at peak performance.
  • Includes syringe, standard needle, and injection cartridge.
  • Contains a 10μL loop and filters.
  • Designed for UPLC system types.
  • Recommended for annual replacement.