Learn More
Waters Corp WATERS Performance Maintenance Kit, ACQUITY i2V Binary Solvent Manager, UPLC, Annual
Catalog No. 50791586 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the ACQUITY i2V Binary Solvent Manager contains all the quality parts necessary to keep the instrument operating at peak performance.
  • Includes tubing (transducer to check valve), an air filter, plungers, flanged seals, and wash seals.
  • Contains a Teflon o-ring, mixer assembly, check valves, and solvent bottle filters.
  • Features an i2V cartridge, filter cartridge, and tubing (SSV to i2V).
  • Designed for UPLC system types.
  • Compatible with ACQUITY Arc Premier System and ACQUITY UPLC H-Class System.
  • Recommended for annual replacement.
  • Comes as 1 unit per package.
Catalog No. 50-791-586 Supplier Waters Corp Supplier No. 201000197
The Performance Maintenance Kit for the ACQUITY i2V Binary Solvent Manager contains all the quality parts necessary to keep the instrument operating at peak performance.
  • Includes tubing (transducer to check valve), an air filter, plungers, flanged seals, and wash seals.
  • Contains a Teflon o-ring, mixer assembly, check valves, and solvent bottle filters.
  • Features an i2V cartridge, filter cartridge, and tubing (SSV to i2V).
  • Designed for UPLC system types.
  • Compatible with ACQUITY Arc Premier System and ACQUITY UPLC H-Class System.
  • Recommended for annual replacement.
  • Comes as 1 unit per package.