Learn More
Waters Corp Waters Rebuild Kit, ACQUITY i2V Primary Pumphead, UPLC System, 1 pk
Catalog No. 502039502 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild an ACQUITY i2V Primary Pumphead.
  • Contains the plunger, seals, and check valve.
  • For ACQUITY i2V Primary Pumphead rebuilds.
  • UPLC system compatible.
  • For ACQUITY UPLC H-Class System and ACQUITY instruments.
  • Replacement as required.
  • For BSM and QSM components.
  • One unit per package.
Catalog No. 50-203-9502 Supplier Waters Corp Supplier No. 700004747
This rebuild kit contains the plunger, seals, and check valve necessary to rebuild an ACQUITY i2V Primary Pumphead.
  • Contains the plunger, seals, and check valve.
  • For ACQUITY i2V Primary Pumphead rebuilds.
  • UPLC system compatible.
  • For ACQUITY UPLC H-Class System and ACQUITY instruments.
  • Replacement as required.
  • For BSM and QSM components.
  • One unit per package.