Learn More
Waters Corp Performance Maintenance Kit, ACQUITY M-Class, Annual, UPLC System
Catalog No. 502391852 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the ACQUITY M-Class Binary and Auxilliary Solvent Managers contains all the necessary quality parts to keep the instrument operating at peak performance on an annual basis.
  • System type: UPLC
  • Instrument: I-Class System
  • Replacement schedule: Annual
  • Component: Column Manager
  • Units per package: 1 pk
  • Kit includes: mixer, plungers, check valves, seals, and filters.
Catalog No. 50-239-1852 Supplier Waters Corp Supplier No. 201000289
Add to Cart
Add to Cart
Request a Quote
The Waters™ Performance Maintenance Kit for the ACQUITY M-Class Binary and Auxilliary Solvent Managers contains all the necessary quality parts to keep the instrument operating at peak performance on an annual basis.
  • System type: UPLC
  • Instrument: I-Class System
  • Replacement schedule: Annual
  • Component: Column Manager
  • Units per package: 1 pk
  • Kit includes: mixer, plungers, check valves, seals, and filters.