Learn More
Waters Corp Waters ACQUITY MS Test Solutions, Qualification, Mass Spectrometry
Catalog No. 50201667 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit offers seven solutions for MS instrument qualification, assessing detector linearity and detection limit.
  • Kit includes two 10 mL vials of 1 mg/mL sulfadimethoxine in methanol.
  • Also 10 mL acetonitrile with 1 mg/mL terfenadine, reserpine, acetaminophen, and caffeine.
  • Includes 10 mL formic acid, a solvent modifier.
  • Sulfadimethoxine dilutions facilitate injector linearity, accuracy, MS detector linearity, system precision, flow rate, carryover, reproducibility, sensitivity, peak width, and signal-to-noise tests.
  • Terfenadine, reserpine, acetaminophen, caffeine solutions combine for gradient testing.
  • Single unit package, 24-month shelf life.
  • Specifications: reversed phase, MS detection, liquid, room temp storage, ambient shipping.
  • Designed for system performance qualification.
  • Hazardous material may incur additional freight charges.
Catalog No. 50-201-667 Supplier Waters Corp Supplier No. 700002728
This kit offers seven solutions for MS instrument qualification, assessing detector linearity and detection limit.
  • Kit includes two 10 mL vials of 1 mg/mL sulfadimethoxine in methanol.
  • Also 10 mL acetonitrile with 1 mg/mL terfenadine, reserpine, acetaminophen, and caffeine.
  • Includes 10 mL formic acid, a solvent modifier.
  • Sulfadimethoxine dilutions facilitate injector linearity, accuracy, MS detector linearity, system precision, flow rate, carryover, reproducibility, sensitivity, peak width, and signal-to-noise tests.
  • Terfenadine, reserpine, acetaminophen, caffeine solutions combine for gradient testing.
  • Single unit package, 24-month shelf life.
  • Specifications: reversed phase, MS detection, liquid, room temp storage, ambient shipping.
  • Designed for system performance qualification.
  • Hazardous material may incur additional freight charges.