Learn More
Waters Corp Waters Corporation Needle Assembly, PEEK, 30 μL
Catalog No. NC9424642 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This PEEK needle assembly is a replacement component designed for use with the ACQUITY UPLC Sample Manager. It features a 30 μL volume and serves as the standard needle assembly for this system.
  • Designed for ACQUITY UPLC Sample Manager
  • Constructed from PEEK material
  • Features a 30 μL volume
  • Recommended for annual replacement
Catalog No. NC9424642 Supplier Waters Corp Supplier No. 700002644
This PEEK needle assembly is a replacement component designed for use with the ACQUITY UPLC Sample Manager. It features a 30 μL volume and serves as the standard needle assembly for this system.
  • Designed for ACQUITY UPLC Sample Manager
  • Constructed from PEEK material
  • Features a 30 μL volume
  • Recommended for annual replacement