Learn More
Waters Corp Waters Performance Maintenance Kit, ACQUITY PDA/TUV 2489/2998, HPLC, UPLC
Catalog No. 50596996 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Waters Performance Maintenance Kit for ACQUITY UPLC TUV/PDA ensures peak instrument performance annually. It facilitates proactive maintenance, guaranteeing analytical accuracy, reliability, and minimizing instrument failures.
  • PerformancePLUS Long Life Lamp: consistent, stable illumination, up to 2,000 hours or one year lifespan.
  • Includes all necessary components for annual upkeep; meets stringent quality standards.
  • Compatible with ACQUITY UPLC PDA, eLambda PDA, TUV, 2489 Dual Wavelength UV, and 2998 PDA detectors.
  • Enhances performance and data accuracy for reliable, reproducible results.
  • Prolongs instrument lifespan by reducing unexpected failures.
  • Recommended for annual preventive maintenance.
  • System compatibility: HPLC, UPLC.
  • Units per package: 1.
Catalog No. 50-596-996 Supplier Waters Corp Supplier No. 201000281
This Waters Performance Maintenance Kit for ACQUITY UPLC TUV/PDA ensures peak instrument performance annually. It facilitates proactive maintenance, guaranteeing analytical accuracy, reliability, and minimizing instrument failures.
  • PerformancePLUS Long Life Lamp: consistent, stable illumination, up to 2,000 hours or one year lifespan.
  • Includes all necessary components for annual upkeep; meets stringent quality standards.
  • Compatible with ACQUITY UPLC PDA, eLambda PDA, TUV, 2489 Dual Wavelength UV, and 2998 PDA detectors.
  • Enhances performance and data accuracy for reliable, reproducible results.
  • Prolongs instrument lifespan by reducing unexpected failures.
  • Recommended for annual preventive maintenance.
  • System compatibility: HPLC, UPLC.
  • Units per package: 1.