Learn More
Waters Corp Tubing Kit, ACQUITY UPLC CM-A, 4 x 50 mm, Stainless Steel
Catalog No. 502390588 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters ACQUITY UPLC CM-A Tubing Kit is designed for ACQUITY H-Class PLUS systems to manage four 50 mm columns. It includes stainless steel tubing and an Active Preheater.
  • Contains stainless steel tubing assemblies to implement CM-A active column managers onto an ACQUITY H-Class PLUS.
  • Supports a four 50 mm column configuration.
  • Can be used to replace all stainless steel tubing in this configuration.
  • Includes the relevant Active Preheater for a PLUS system.
Catalog No. 50-239-0588 Supplier Waters Corp Supplier No. 205001832
Add to Cart
Add to Cart
The Waters ACQUITY UPLC CM-A Tubing Kit is designed for ACQUITY H-Class PLUS systems to manage four 50 mm columns. It includes stainless steel tubing and an Active Preheater.
  • Contains stainless steel tubing assemblies to implement CM-A active column managers onto an ACQUITY H-Class PLUS.
  • Supports a four 50 mm column configuration.
  • Can be used to replace all stainless steel tubing in this configuration.
  • Includes the relevant Active Preheater for a PLUS system.