Learn More
Waters Corp Waters ACQUITY Premier Protein SEC Column, 250 Å, 1.7 μm, 4.6 x 300 mm
Catalog No. NC2067197 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY Premier Protein SEC 250 Å, 1.7 μm Columns separate protein size variants (10,000 to 650,000 Daltons). They combine MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to minimize unwanted interactions, reducing method development time. Designed for UPLC SEC, each includes a Waters mAb Size Variant Standard (p/n: 186009429).
  • Enables reliable protein size variant separations (10,000 to 650,000 Daltons).
  • Uses MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Minimizes secondary ionic or hydrophobic interactions.
  • Reduces method development needs through “generic” or “platform-type” methods.
  • Optimized for UPLC SEC applications with low LC system volume.
This item is not returnable. View return policy
Catalog No. NC2067197 Supplier Waters Corp Supplier No. 176005072
Add to Cart
Add to Cart
Request a Quote
ACQUITY Premier Protein SEC 250 Å, 1.7 μm Columns separate protein size variants (10,000 to 650,000 Daltons). They combine MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies to minimize unwanted interactions, reducing method development time. Designed for UPLC SEC, each includes a Waters mAb Size Variant Standard (p/n: 186009429).
  • Enables reliable protein size variant separations (10,000 to 650,000 Daltons).
  • Uses MaxPeak Premier High Performance Surfaces and BEH-PEO Particle technologies.
  • Minimizes secondary ionic or hydrophobic interactions.
  • Reduces method development needs through “generic” or “platform-type” methods.
  • Optimized for UPLC SEC applications with low LC system volume.