Learn More
Waters Corp Waters Corporation Column, BEH Amide, 1.7 μm, 2.1 mm X 100 mm
Supplier:  Waters Corp 186009505
Encompass_Preferred
Designed to retain extremely polar compounds like carbohydrates and sugars, these columns utilize BEH technology and a stable amide phase (pH 2 to 11). MaxPeak High Performance Surface hardware reduces analyte interactions, improving peak shape and signal intensity.
  • Retains extremely polar compounds.
  • Chemically stable, trifunctionally bonded amide phase.
  • pH stable from 2 to 11.
  • MaxPeak High Performance Surface hardware.
  • Reduces unwanted analyte/surface interactions.
  • Hydrophilic Interaction (HILIC) separation mode.
  • Hybrid, spherical particle substrate with low silanol activity.
  • Parker-style endfitting.
  • 1.7 µm particle size, 130 Å pore size, 185 surface area.
  • Compatible with UHPLC and UPLC systems (LC technique).
Catalog No. 50-222-4221