Learn More
Waters Corp ACQUITY Method Validation Kit, BEH C18, 130Å, 1.7 μm, 2.1 x 150 mm
Catalog No. 502391336 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three columns for method development, offering high-performance separations. It features MaxPeak High Performance Surface hardware to reduce unwanted analyte/surface interactions and improve peak shape. The BEH particle provides a wide usable pH range (pH 1–12). The VanGuard Fit design protects from contaminants and extends lifetime.
  • Method validation kit includes three columns packed with different material lots.
  • Offers a universal column choice for high-performance separations.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, improving peak shape and signal.
  • Trifunctionally bonded BEH particle provides a wide usable pH range (pH 1–12), ideal for method development.
  • VanGuard Fit column design protects from contaminants and extends lifetime.
Catalog No. 50-239-1336 Supplier Waters Corp Supplier No. 186009625
Add to Cart
Add to Cart
This method validation kit includes three columns for method development, offering high-performance separations. It features MaxPeak High Performance Surface hardware to reduce unwanted analyte/surface interactions and improve peak shape. The BEH particle provides a wide usable pH range (pH 1–12). The VanGuard Fit design protects from contaminants and extends lifetime.
  • Method validation kit includes three columns packed with different material lots.
  • Offers a universal column choice for high-performance separations.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, improving peak shape and signal.
  • Trifunctionally bonded BEH particle provides a wide usable pH range (pH 1–12), ideal for method development.
  • VanGuard Fit column design protects from contaminants and extends lifetime.