Learn More
Waters Corp ACQUITY Premier BEH C18 Column, 130Å, 1.7 μm
Catalog No. 502392320 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit provides three columns, each packed with a unique lot. It serves as a universal column for high-performance separations and method development. MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. The trifunctionally bonded BEH particle offers a wide usable pH range (pH 1 to 12), ideal for method development. The VanGuard Fit design protects the column from contaminants and extends its lifetime.
  • Universal column choice for high-performance separations.
  • Hardware technology reduces unwanted analyte/surface interactions, minimizing poor peak shape and signal intensity losses.
  • Trifunctionally bonded particle offers a wide usable pH range (pH 1 to 12), ideal for method development.
  • Column design protects from contaminants and increases lifetime.
Catalog No. 50-239-2320 Supplier Waters Corp Supplier No. 186009624
This method validation kit provides three columns, each packed with a unique lot. It serves as a universal column for high-performance separations and method development. MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. The trifunctionally bonded BEH particle offers a wide usable pH range (pH 1 to 12), ideal for method development. The VanGuard Fit design protects the column from contaminants and extends its lifetime.
  • Universal column choice for high-performance separations.
  • Hardware technology reduces unwanted analyte/surface interactions, minimizing poor peak shape and signal intensity losses.
  • Trifunctionally bonded particle offers a wide usable pH range (pH 1 to 12), ideal for method development.
  • Column design protects from contaminants and increases lifetime.