Learn More
Waters Corp Waters ACQUITY Premier BEH C18 Column, 1.7 μm, 2.1 x 50 mm, MVK
Catalog No. 502039105 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier BEH C18 columns are designed for high-performance liquid chromatography and are a universal choice for method development. This method validation kit (MVK) includes three columns, each from a different material lot. MaxPeak High Performance Surface technology minimizes unwanted analyte/surface interactions, enhancing peak shape and signal intensity. The trifunctionally bonded BEH particle enables a wide usable pH range of 1 to 12.
  • Method validation kit (MVK) includes three columns, each from a different material lot.
  • Universal column choice for high performance separations.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Trifunctionally bonded BEH particle provides a wide usable pH range (pH 1 to 12).
Catalog No. 50-203-9105 Supplier Waters Corp Supplier No. 186009620
The ACQUITY Premier BEH C18 columns are designed for high-performance liquid chromatography and are a universal choice for method development. This method validation kit (MVK) includes three columns, each from a different material lot. MaxPeak High Performance Surface technology minimizes unwanted analyte/surface interactions, enhancing peak shape and signal intensity. The trifunctionally bonded BEH particle enables a wide usable pH range of 1 to 12.
  • Method validation kit (MVK) includes three columns, each from a different material lot.
  • Universal column choice for high performance separations.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Trifunctionally bonded BEH particle provides a wide usable pH range (pH 1 to 12).