Learn More
Waters Corp Waters Column, C18, 1.7 μm, 2.1 mm, 150 mm
Catalog No. 502039061 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for high-performance separations and method development. It utilizes MaxPeak High Performance Surface hardware technology to minimize unwanted analyte/surface interactions, improving peak shape and signal intensity. Its trifunctionally bonded BEH particle offers a wide usable pH range (1 to 12 pH).
  • Features MaxPeak High Performance Surface hardware to reduce analyte/surface interactions.
  • Provides wide usable pH range (1 to 12 pH) for method development.
  • Offers UPLC separation performance for diverse analytes with high efficiency.
  • Enhances chemical stability and maintains excellent peak shape for basic analytes.
  • Optional VanGuard FIT technology protects against fouling.
Catalog No. 50-203-9061 Supplier Waters Corp Supplier No. 186009454
This column is designed for high-performance separations and method development. It utilizes MaxPeak High Performance Surface hardware technology to minimize unwanted analyte/surface interactions, improving peak shape and signal intensity. Its trifunctionally bonded BEH particle offers a wide usable pH range (1 to 12 pH).
  • Features MaxPeak High Performance Surface hardware to reduce analyte/surface interactions.
  • Provides wide usable pH range (1 to 12 pH) for method development.
  • Offers UPLC separation performance for diverse analytes with high efficiency.
  • Enhances chemical stability and maintains excellent peak shape for basic analytes.
  • Optional VanGuard FIT technology protects against fouling.