Learn More
Waters Corp Waters ACQUITY Premier BEH C18 Column, 1.7 μm, 2.1 mm, 50 mm
Catalog No. 502039059 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY Premier BEH C18 Column provides high-performance separations and method development. It features MaxPeak High Performance Surface hardware to reduce analyte/surface interactions, ensuring optimal peak shape and signal. The trifunctionally bonded BEH particle offers a wide usable pH range (pH 1–12), delivering efficiency, peak symmetry, and chemical stability. Produced in a cGMP, ISO 9001 certified plant.
  • MaxPeak High Performance Surface hardware technology reduces analyte/surface interactions.
  • Trifunctionally bonded BEH particle offers a wide usable pH range (pH 1–12).
  • Designed for high-performance separations and method development.
  • Superior low pH stability and ultra-low column bleed for high-sensitivity MS applications.
  • Engineered for use with UPLC systems; produced in a cGMP, ISO 9001 certified plant.
Catalog No. 50-203-9059 Supplier Waters Corp Supplier No. 186009452
The ACQUITY Premier BEH C18 Column provides high-performance separations and method development. It features MaxPeak High Performance Surface hardware to reduce analyte/surface interactions, ensuring optimal peak shape and signal. The trifunctionally bonded BEH particle offers a wide usable pH range (pH 1–12), delivering efficiency, peak symmetry, and chemical stability. Produced in a cGMP, ISO 9001 certified plant.
  • MaxPeak High Performance Surface hardware technology reduces analyte/surface interactions.
  • Trifunctionally bonded BEH particle offers a wide usable pH range (pH 1–12).
  • Designed for high-performance separations and method development.
  • Superior low pH stability and ultra-low column bleed for high-sensitivity MS applications.
  • Engineered for use with UPLC systems; produced in a cGMP, ISO 9001 certified plant.